Methyl-5-bromo-2-iodobenzoate ≥95.0% (by GC)

Supplier: TCI America

M3084-1G M3084-5G
TCM3084-1GEA 28.44 CAD
TCM3084-1G TCM3084-5G
Methyl-5-bromo-2-iodobenzoate ≥95.0% (by GC)
Methyl 5-bromo-2-iodobenzoate
Formula: C₈H₆BrIO₂
Boiling Pt: 134°C
MDL Number: MFCD00144771
CAS Number: 181765-86-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 43.0 to 47.0 deg-C
Acid Value max. 3.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR