4-(Diethylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-(Diethylcarbamoyl)benzeneboronic acid, 4-(N,N-Diethylaminocarbonyl)phenylboronic acid

D4681-1G D4681-5G
TCD4681-1GEA 200.66 CAD
TCD4681-1G TCD4681-5G
4-(Diethylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Diethylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
MDL Number: MFCD03411949
CAS Number: 389621-80-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR