(4-Bromo-2-nitrophenyl)acetic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Bromo-2-nitrophenylacetic acid
B5159-1G
B5159-5G
TCB5159-1GEA
164.32
CAD
TCB5159-1G
TCB5159-5G
(4-Bromo-2-nitrophenyl)acetic acid ≥97.0% (by titrimetric analysis)
(4-Bromo-2-nitrophenyl)acetic acid
Formula:
C₈H₆BrNO₄ MW: 260.0400 g/mol |
MDL Number:
MFCD05663950 CAS Number: 6127-11-3 |
Specification Test Results
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...