Methyl-2-aminothiazole-4-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2581-1G
M2581-5G
TCM2581-1GEA
120.08
CAD
TCM2581-1G
TCM2581-5G
Methyl-2-aminothiazole-4-carboxylate ≥98.0% (by GC, titration analysis)
Methyl 2-aminothiazole-4-carboxylate
Formula:
C₅H₆N₂O₂S |
MDL Number:
MFCD00622441 CAS Number: 118452-04-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 171.0 to 176.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...