5-(5-Fluoro-2-hydroxyphenyl)isoxazole ≥98.0% (by GC)

Supplier: TCI America
Warning

F1049-1G F1049-5G
TCF1049-1GEA 101.12 CAD
TCF1049-1G TCF1049-5G
5-(5-Fluoro-2-hydroxyphenyl)isoxazole ≥98.0% (by GC)
5-(5-Fluoro-2-hydroxyphenyl)isoxazole
Formula: C₉H₆FNO₂
MDL Number: MFCD03070563
CAS Number: 288401-62-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR