3-Iodo-6-nitro-1H-indazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Iodo-6-nitroindazole

I1004-1G I1004-5G
TCI1004-1GEA 194.34 CAD
TCI1004-1G TCI1004-5G
3-Iodo-6-nitro-1H-indazole ≥98.0% (by GC)
3-Iodo-6-nitro-1H-indazole
Formula: C₇H₄IN₃O₂
MDL Number: MFCD07781652
CAS Number: 70315-70-7

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR