Methyl-1H-indazole-6-carboxylate ≥97.0% (by GC)

Supplier: TCI America

M2823-1G M2823-5G
TCM2823-1GEA 47.4 CAD
TCM2823-1G TCM2823-5G
Methyl-1H-indazole-6-carboxylate ≥97.0% (by GC)
Methyl 1H-indazole-6-carboxylate
Formula: C₉H₈N₂O₂
MDL Number: MFCD07371612
CAS Number: 170487-40-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR