Ethyl-1H-indazole-5-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl Indazole-5-carboxylate, Ethyl 1H-indazole-5-carboxylate, Indazole-5-carboxylic acid ethyl ester

E1067-1G E1067-5G
TCE1067-1GEA 200.66 CAD
TCE1067-1G TCE1067-5G
Ethyl-1H-indazole-5-carboxylate ≥98.0% (by GC)
Ethyl 1H-indazole-5-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD03426220
CAS Number: 192944-51-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR