2-Bromoquinoxaline ≥98.0% (by GC)

Supplier: TCI America
Warning

B4632-1G B4632-200MG
TCB4632-1GEA 199.16 CAD
TCB4632-1G TCB4632-200MG
2-Bromoquinoxaline ≥98.0% (by GC)
2-Bromoquinoxaline
Formula: C₈H₅BrN₂
MDL Number: MFCD07368206
CAS Number: 36856-91-4

Order Now

Specification Test Results

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR