2-(4-Bromophenyl)imidazo[1,2-a]pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4871-1G
B4871-5G
TCB4871-1GEA
153.73
CAD
TCB4871-1G
TCB4871-5G
2-(4-Bromophenyl)imidazo[1,2-a]pyridine ≥98.0% (by GC, titration analysis)
2-(4-Bromophenyl)imidazo[1,2-a]pyridine
Formula:
C₁₃H₉BrN₂ |
MDL Number:
MFCD00218250 CAS Number: 34658-66-7 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 215.0 to 219.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...