2-Aminoisonicotinamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-Aminopyridine-4-carboxamide, 2-Amino-4-pyridinecarboxamide

A2899-1G A2899-5G
TCA2899-1GEA 77.42 CAD
TCA2899-1G TCA2899-5G
2-Aminoisonicotinamide ≥98.0% (by HPLC, titration analysis)
2-Aminoisonicotinamide
Formula: C₆H₇N₃O
MW: 137.14 g/mol
MDL Number: MFCD03791262
CAS Number: 13538-42-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR