4-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
M3057-1G
M3057-5G
TCM3057-1GEA
33.18
CAD
TCM3057-1G
TCM3057-5G
4-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula:
C₁₄H₁₉BO₄ MW: 262.11 g/mol |
MDL Number:
MFCD02179438 CAS Number: 171364-80-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...