3-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2665-1G M2665-5G
TCM2665-1GEA 61.62 CAD
TCM2665-1G TCM2665-5G
3-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MDL Number: MFCD03789258
CAS Number: 480425-35-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR