Cyclohexylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C3101-1G C3101-5G
TCC3101-1GEA 49.61 CAD
TCC3101-1G TCC3101-5G
Cyclohexylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
Cyclohexylboronic acid pinacol ester
Formula: C₁₂H₂₃BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD04038749
CAS Number: 87100-15-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR