2,4-Dibromobenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America
Warning

D5002-1G D5002-5G
76219-116EA 86.9 CAD
76219-116 76219-118
2,4-Dibromobenzyl alcohol ≥98.0% (by GC)
2,4-Dibromobenzyl alcohol
Formula: C₇H₆Br₂O
MDL Number: MFCD09996906
CAS Number: 666747-06-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C
Solubility in N,N-DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR