4-tert-Butylphenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 4-tert-Butylphenylisothiocyanate

B4984-1G B4984-5G
TCB4984-1GEA 49.61 CAD
TCB4984-1G TCB4984-5G
4-tert-Butylphenyl isothiocyanate ≥98.0% (by GC)
4-tert-Butylphenyl isothiocyanate
Formula: C₁₁H₁₃NS
MW: 191.2926 g/mol
Boiling Pt: 277°C
Storage Temperature: Refrigerator
MDL Number: MFCD00022059
CAS Number: 19241-24-8
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR