4-Fluoro-1-naphthaleneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

F1080-1G F1080-200MG
TCF1080-1GEA 222.78 CAD
TCF1080-1G TCF1080-200MG
4-Fluoro-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
4-Fluoro-1-naphthaleneboronic acid
Formula: C₁₀H₇BBrFO₂
MDL Number: MFCD04039221
CAS Number: 182344-25-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR