2-Naphthyl boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
T3298-1G
T3298-5G
TCT3298-1GEA
44.24
CAD
TCT3298-1G
TCT3298-5G
2-Naphthyl boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Naphthyl boronic acid pinacol ester
Formula:
C₁₆H₁₉BO₂ |
MDL Number:
MFCD05663868 CAS Number: 256652-04-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...