1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1,3,5-Trimethylpyrazole-4-boronic acid Pinacol ester, 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1,3,5-Trimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3409-1G T3409-200MG
76220-620EA 186.44 CAD
76220-620 76220-622
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₂H₂₁BN₂O₂
MW: 236.12 g/mol
MDL Number: MFCD06659062
CAS Number: 844891-04-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR