6-Chloroimidazo[1,2-a]pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C3370-1G
C3370-5G
TCC3370-1GEA
56.88
CAD
TCC3370-1G
TCC3370-5G
6-Chloroimidazo[1,2-a]pyridine ≥98.0% (by GC, titration analysis)
6-Chloroimidazo[1,2-a]pyridine
Formula:
C₇H₅ClN₂ Boiling Pt: 132°C |
MDL Number:
MFCD05663807 CAS Number: 6188-25-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...