6-Chloroimidazo[1,2-b]pyridazine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C3127-1G
C3127-5G
TCC3127-1GEA
37.92
CAD
TCC3127-1G
TCC3127-5G
6-Chloroimidazo[1,2-b]pyridazine ≥98.0% (by GC, titration analysis)
6-Chloroimidazo[1,2-b]pyridazine
Formula:
C₆H₄ClN₃ MW: 153.5691 g/mol |
MDL Number:
MFCD07778345 CAS Number: 6775-78-6 |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...