4-Bromo-2-nitrobenzaldehyde ≥96.0% (by GC)

Supplier: TCI America
Warning

B4146-1G B4146-5G
TCB4146-1GEA 52.14 CAD
TCB4146-1G TCB4146-5G
4-Bromo-2-nitrobenzaldehyde ≥96.0% (by GC)
4-Bromo-2-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
MDL Number: MFCD00463687
CAS Number: 5551-12-2

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR