3-(4-Methoxyphenyl)-1-phenyl-1H-pyrazol-5-amine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Amino-3-(4-methoxyphenyl)-1-phenylpyrazole

A3055-1G A3055-5G
TCA3055-1GEA 165.9 CAD
TCA3055-1G TCA3055-5G
3-(4-Methoxyphenyl)-1-phenyl-1H-pyrazol-5-amine ≥98.0% (by HPLC, titration analysis)
3-(4-Methoxyphenyl)-1-phenyl-1H-pyrazol-5-amine
Formula: C₁₆H₁₅N₃O
MDL Number: MFCD00197061
CAS Number: 19652-13-2

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR