4-(3-Pyridyl)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

P2332-1G P2332-200MG
TCP2332-1GEA 124.82 CAD
TCP2332-1G TCP2332-200MG
4-(3-Pyridyl)benzaldehyde ≥98.0% (by GC)
4-(3-Pyridyl)benzaldehyde
Formula: C₁₂H₉NO
MDL Number: MFCD01631910
CAS Number: 127406-55-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR