(R)-(+)-Pulegone ≥85.0% (by GC)

Supplier: TCI America
Warning

P2331-25ML P2331-5ML
TCP2331-25MLEA 74.26 CAD
TCP2331-25ML TCP2331-5ML
(R)-(+)-Pulegone ≥85.0% (by GC)
(R)-(+)-Pulegone
Formula: C₁₀H₁₆O
Boiling Pt: 217°C
Flash Pt: 82°C
MDL Number: MFCD00063000
CAS Number: 89-82-7
EINECS: 201-943-2
Merck Index: 14,07937

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 85.0 %
Specific rotation [a]20/D +22.0 to +27.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR