2,4-Dichlorobenzyl bromide ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
D4475-1G
D4475-5G
TCD4475-1GEA
69.52
CAD
TCD4475-1G
TCD4475-5G
2,4-Dichlorobenzyl bromide ≥98.0% (by GC)
2,4-Dichlorobenzyl bromide
Formula:
C₇H₅BrCl₂ Boiling Pt: 112°C Storage Temperature: Refrigerator |
MDL Number:
MFCD03208523 CAS Number: 20443-99-6 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 30.0 to 34.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...