2-Chloro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C3222-1G C3222-200MG
TCC3222-1GEA 221.2 CAD
TCC3222-1G TCC3222-200MG
2-Chloro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-4-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
MDL Number: MFCD02684318
CAS Number: 313545-44-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR