5-Bromo-2-chlorobenzaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4756-1G
B4756-5G
TCB4756-1GEA
39.5
CAD
TCB4756-1G
TCB4756-5G
5-Bromo-2-chlorobenzaldehyde ≥98.0% (by GC)
5-Bromo-2-chlorobenzaldehyde
Formula:
C₇H₄BrClO |
MDL Number:
MFCD08445659 CAS Number: 189628-37-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...