5'-Bromo-2'-hydroxypropiophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(5-Bromo-2-hydroxyphenyl)propan-1-one

B4769-1G B4769-5G
TCB4769-1GEA 39.5 CAD
TCB4769-1G TCB4769-5G
5'-Bromo-2'-hydroxypropiophenone ≥98.0% (by GC, titration analysis)
5'-Bromo-2'-hydroxypropiophenone
Formula: C₉H₉BrO₂
Boiling Pt: 156°C
MDL Number: MFCD00218577
CAS Number: 17764-93-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR