Tris[3,5-bis(trifluoromethyl)phenyl]phosphine ≥98.0% (by GC)

Supplier: TCI America
Warning

T2526-1G T2526-5G
TCT2526-1GEA 319.16 CAD
TCT2526-1G TCT2526-5G
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine ≥98.0% (by GC)
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Formula: C₂₄H₉F₁₈P
Boiling Pt: 130°C
MDL Number: MFCD00042428
CAS Number: 175136-62-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR