5-Nitro-2,4-xylidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,4-Dimethyl-5-nitroaniline, 5-Nitro-2,4-xylidine

D5196-1G D5196-5G
76219-230EA 109.02 CAD
76219-230 76219-232
5-Nitro-2,4-xylidine ≥98.0% (by GC, titration analysis)
5-Nitro-2,4-xylidine
Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
MDL Number: MFCD00025199
CAS Number: 2124-47-2

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR