4′-Hydroxy-2-phenylacetophenone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4722-1G
B4722-5G
TCB4722-1GEA
123.24
CAD
TCB4722-1G
TCB4722-5G
4′-Hydroxy-2-phenylacetophenone ≥98.0% (by GC, titration analysis)
4′-Hydroxy-2-phenylacetophenone
Formula:
C₁₄H₁₂O₂ |
MDL Number:
MFCD00002360 CAS Number: 2491-32-9 EINECS: 219-654-5 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...