4-Benzyloxybenzophenone ≥98.0% (by GC)

Supplier: TCI America

B4504-25G B4504-5G
TCB4504-25GEA 330.54 CAD
TCB4504-25G TCB4504-5G
4-Benzyloxybenzophenone ≥98.0% (by GC)
4-Benzyloxybenzophenone
Formula: C₁₄H₉F₃O
MDL Number: MFCD00017508
CAS Number: 54589-41-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR