Benzyl-4-chlorophenyl ketone ≥98.0% (by GC)
Supplier: TCI America
B4704-25G
B4704-5G
TCB4704-25GEA
189.6
CAD
TCB4704-25G
TCB4704-5G
Benzyl-4-chlorophenyl ketone ≥98.0% (by GC)
Benzyl 4-chlorophenyl ketone
Formula:
C₁₄H₁₁ClO Boiling Pt: 187°C |
MDL Number:
MFCD00016342 CAS Number: 1889-71-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...