1-(2-Tetrahydropyranyl)-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole

T3408-1G T3408-5G
76220-616EA 45.82 CAD
76220-616 76220-618
1-(2-Tetrahydropyranyl)-1H-pyrazole ≥98.0% (by GC)
1-(2-Tetrahydropyranyl)-1H-pyrazole
Formula: C₈H₁₂N₂O
MW: 152.20 g/mol
Boiling Pt: 65°C
Flash Pt: 104°C
MDL Number: MFCD11044692
CAS Number: 449758-17-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR