4-Bromo-4'-chlorobenzophenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (4-Bromophenyl)(4-chlorophenyl)methanone

B4224-1G B4224-5G
TCB4224-1GEA 237 CAD
TCB4224-1G TCB4224-5G
4-Bromo-4'-chlorobenzophenone ≥98.0% (by GC)
4-Bromo-4'-chlorobenzophenone
Formula: C₁₃H₈BrClO
MW: 295.5590 g/mol
MDL Number: MFCD00045208
CAS Number: 27428-57-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR