2-Amino-4'-bromoacetophenone hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
A3030-1G
A3030-5G
TCA3030-1GEA
104.28
CAD
TCA3030-1G
TCA3030-5G
2-Amino-4'-bromoacetophenone hydrochloride ≥98.0% (by HPLC, total nitrogen)
2-Amino-4'-bromoacetophenone hydrochloride
Formula:
C₈H₉BrClNO Melting Pt: 285°C |
MDL Number:
MFCD00051998 CAS Number: 5467-72-1 EINECS: 226-778-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...