3-Fluoro-4-formylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
F1051-1G
F1051-5G
TCF1051-1GEA
63.2
CAD
TCF1051-1G
TCF1051-5G
3-Fluoro-4-formylphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-4-formylphenylboronic acid
Formula:
C₇H₆BFO₃ |
MDL Number:
MFCD02093051 CAS Number: 248270-25-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...