2-Amino-5-pyrimidineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3087-1G A3087-5G
TCA3087-1GEA 139.04 CAD
TCA3087-1G TCA3087-5G
2-Amino-5-pyrimidineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Amino-5-pyrimidineboronic acid pinacol ester
Formula: C₁₀H₁₆BN₃O₂
MDL Number: MFCD06795685
CAS Number: 402960-38-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 210.0 to 214.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR