5-Iodo-1-methyl-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
I0963-1G
I0963-5G
TCI0963-1GEA
183.28
CAD
TCI0963-1G
TCI0963-5G
5-Iodo-1-methyl-1H-pyrazole ≥98.0% (by GC)
5-Iodo-1-methyl-1H-pyrazole
Formula:
C₄H₅IN₂ |
MDL Number:
MFCD11109327 CAS Number: 34091-51-5 |
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...