4-Bromo-1-ethyl-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5100-1G B5100-5G
TCB5100-1GEA 109.02 CAD
TCB5100-1G TCB5100-5G
4-Bromo-1-ethyl-1H-pyrazole ≥98.0% (by GC)
4-Bromo-1-ethyl-1H-pyrazole
Formula: C₅H₇BrN₂
Boiling Pt: 108°C
MDL Number: MFCD08235252
CAS Number: 71229-85-1

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR