2-Hydroxy-4-methylbenzophenone ≥97.0% (by GC)

Supplier: TCI America
Warning

H1575-25G H1575-5G
TCH1575-25GEA 257.54 CAD
TCH1575-25G TCH1575-5G
2-Hydroxy-4-methylbenzophenone ≥97.0% (by GC)
2-Hydroxy-4-methylbenzophenone
Formula: C₁₄H₁₂O₂
Boiling Pt: 234°C
MDL Number: MFCD01099035
CAS Number: 3098-18-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR