2,3,3,5-Tetramethylindolenine ≥98.0% (by GC)

Supplier: TCI America
Warning

T3458-25G T3458-5G
76220-658EA 320.74 CAD
76220-658 76220-660
2,3,3,5-Tetramethylindolenine ≥98.0% (by GC)
2,3,3,5-Tetramethylindolenine
Formula: C₁₂H₁₅N
Boiling Pt: 125°C
MDL Number: MFCD03093020
CAS Number: 25981-82-2

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR