3-Formyl-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
F1088-1G
F1088-5G
TCF1088-1GEA
132.72
CAD
TCF1088-1G
TCF1088-5G
3-Formyl-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Formyl-2-methoxyphenylboronic acid
Formula:
C₈H₉BO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD08689486 CAS Number: 480424-49-5 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...