4-Chloro-3-nitrophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C3212-1G C3212-5G
TCC3212-1GEA 172.22 CAD
TCC3212-1G TCC3212-5G
4-Chloro-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-3-nitrophenylboronic acid
Formula: C₆H₅BClNO₄
MDL Number: MFCD02258950
CAS Number: 151169-67-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR