1,4-Dibromo-3,5-difluorobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D5261-1G D5261-5G
76219-258EA 67.94 CAD
76219-258 76219-260
1,4-Dibromo-3,5-difluorobenzene ≥98.0% (by GC)
1,4-Dibromo-3,5-difluorobenzene
Formula: C₆H₂Br₂F₂
Storage Temperature: Refrigerator
MDL Number: MFCD07783665
CAS Number: 128259-71-2

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR