1,4-Dibromo-2,3-difluorobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4983-1G D4983-5G
TCD4983-1GEA 71.1 CAD
TCD4983-1G TCD4983-5G
1,4-Dibromo-2,3-difluorobenzene ≥98.0% (by GC)
1,4-Dibromo-2,3-difluorobenzene
Formula: C₆H₂Br₂F₂
Storage Temperature: Refrigerator
MDL Number: MFCD09835192
CAS Number: 156682-52-9

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR