2-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
C3213-1G
C3213-200MG
TCC3213-1GEA
455.04
CAD
TCC3213-1G
TCC3213-200MG
2-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-3-(trifluoromethyl)phenylboronic acid
Formula:
C₇H₅BClF₃O₂ MW: 224.3726 g/mol |
MDL Number:
MFCD09258745 CAS Number: 957061-11-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...