3',5'-Dichloro-2,2,2-trifluoroacetophenone ≥96.0% (by GC)

Supplier: TCI America
Warning

D5113-1G D5113-5G
TCD5113-1GEA 69.52 CAD
TCD5113-1G TCD5113-5G
3',5'-Dichloro-2,2,2-trifluoroacetophenone ≥96.0% (by GC)
3',5'-Dichloro-2,2,2-trifluoroacetophenone
Formula: C₈H₃Cl₂F₃O
Boiling Pt: 87°C
MDL Number: MFCD01319996
CAS Number: 130336-16-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR