4'-Bromo-2'-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

B5236-1G B5236-5G
TCB5236-1GEA 107.44 CAD
TCB5236-1G TCB5236-5G
4'-Bromo-2'-fluoroacetophenone ≥98.0% (by GC)
4'-Bromo-2'-fluoroacetophenone
Formula: C₈H₆BrFO
MDL Number: MFCD03411551
CAS Number: 625446-22-2

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR